This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TPBPA SIRNA Assays ›
Gene Symbol:
Tpbpa
Gene Name:
trophoblast specific protein alpha
Gene Aliases:
4311, AV033951, Tb-1, Tb1, Tpbp, b2b1247Clo
Chromosome Location:
Chr.13: 60938692 - 60941935 on Build GRCm38
UniGene ID:
Mm.2372
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS247943
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tpbpa
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009411.4 NP_033437.2 2, 3 186
GenBank mRNA AK005406.1 - 2, 3 186
AK005417.1 - 2, 3 187
AK005438.1 - 2, 3 186
AK005446.1 - 2, 3 187
AK005454.1 - 2, 3 188
AK005499.1 - 2, 3 187
AK011401.1 - 2, 3 187
AK014443.1 - 2, 3 188
AK018797.1 - 2, 3 187
AK018800.1 - 2, 3 186
AK018802.1 - 2, 3 186
AK018803.1 - 2, 3 187
AK018806.1 - 2, 3 187
AK018807.1 - 2, 3 187
AK018811.1 - 2, 3 187
AK018812.1 - 2, 3 187
AK018813.1 - 2, 3 187
AK018814.1 - 2, 3 187
AK018815.1 - 2, 3 186
AK018819.1 - 2, 3 187
AK018821.1 - 2, 3 187
AK018822.1 - 2, 3 188
AK018823.1 - 2, 3 187
AK018825.1 - 2, 3 188
AK018826.1 - 2, 3 187
AK028071.1 - 2, 3 186
AK160140.1 - 2, 3 180
AY034575.1 - 2, 3 162
BC099391.1 - 2, 3 173 30787380
X17071.1 - 2, 3 178

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

JNK cascade
proteolysis involved in cellular protein catabolic process
cellular response to organic cyclic compound
molecular_function
cysteine-type endopeptidase activity
cysteine-type peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header