This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DESI2 SIRNA Assays ›
Gene Symbol:
Desi2 Gm16432
Gene Name:
desumoylating isopeptidase 2
predicted gene 16432
Gene Aliases:
5830417C01Rik, DeSI-2, Fam152a, Pppde1
EG545391, EG666250
Chromosome Location:
Chr.1: 178187231 - 178252598 on Build GRCm38
Chromosome Location:
Chr.1: 177991435 - 178048291 on Build GRCm38
UniGene ID:
Mm.440132
Mm.442974
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID MSS246565
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Desi2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024282.3 NP_077244.1 1 514
GenBank mRNA AK020014.1 - 1 399
AK029574.1 - 1 515
AK049579.1 - 1 283
AK146492.1 - 1 247
AK160733.1 - 1 103
BC002200.1 - 1 183 3489351
BC046816.1 - 1 221 5702476
Gene Symbol: Gm16432
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK139440.1 - 256

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016076, AF229834, AF151904, BC004485, BI459676, and others.
This siRNA also targets the following transcripts in Rat: NM_001013873, XM_006250317, XM_008769785, BC083584, FQ226068.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0