This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FCHO1 SIRNA Assays ›
Gene Symbol:
Fcho1
Gene Name:
FCH domain only 1
Gene Aliases:
3322402E17Rik, N28152
Chromosome Location:
Chr.8: 71708387 - 71725704 on Build GRCm38
UniGene ID:
Mm.22025
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS246087
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fcho1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028715.3 NP_082991.3 6 439
GenBank mRNA AK014382.1 - 6 424
AK088957.1 - 6 423
AK137333.1 - 6 426
AK172927.1 - 3 310
BC032207.1 - 5 415 5323527

Back To Top

More Information


Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
clathrin coat assembly
clathrin-mediated endocytosis
AP-2 adaptor complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0