This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHKG2 SIRNA Assays ›
Gene Symbol:
Phkg2 Gm166
Gene Name:
phosphorylase kinase, gamma 2 (testis)
predicted gene 166
Gene Aliases:
1500017I02Rik
-
Chromosome Location:
Chr.7: 127573344 - 127583307 on Build GRCm38
Chromosome Location:
Chr.7: 127582383 - 127588609 on Build GRCm38
UniGene ID:
Mm.274473
Mm.25220
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID MSS245316
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phkg2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026888.3 NP_081164.2 10 1123
GenBank mRNA AA198694.1 - 5 537
AK005277.1 - 10 1068
BC138913.1 - 10 1037 8861935
BC145734.1 - 10 1037 40131037
BQ176174.1 - 1 518
Gene Symbol: Gm166
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033040.3 NP_001028212.1 9 2111
GenBank mRNA BC066021.1 - 9 1692 5717506

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycogen metabolic process
glycogen biosynthetic process
protein phosphorylation
phosphorylation
biological_process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
phosphorylase kinase activity
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0