This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MARCH11 SIRNA Assays ›
Gene Symbol:
March11
Gene Name:
membrane-associated ring finger (C3HC4) 11
Gene Aliases:
9630025C22
Chromosome Location:
Chr.15: 26308904 - 26409594 on Build GRCm38
UniGene ID:
Mm.54257
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS242319
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: March11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177597.5 NP_808265.2 3 907

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001102562, BC150513, BC150514.
This siRNA also targets the following transcripts in Rat: NM_001101828, AB048841.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0