This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADRBK2 SIRNA Assays ›
Gene Symbol:
Adrbk2
Gene Name:
adrenergic receptor kinase, beta 2
Gene Aliases:
4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3
Chromosome Location:
Chr.5: 112910478 - 113015951 on Build GRCm38
UniGene ID:
Mm.285619
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS241492
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adrbk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285806.1 NP_001272735.1 4 390
NM_177078.4 NP_796052.2 4 351
GenBank mRNA AK048763.1 - 4 316
AK076533.1 - 4 249
AK134672.1 - 4 328
AK141186.1 - 4 368
AK172513.1 - 4 303
CJ104587.1 - 4 352

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
intracellular protein transport
signal transduction
phosphorylation
receptor internalization
termination of G-protein coupled receptor signaling pathway
inositol phosphate metabolic process
rhodopsin metabolic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
D1 dopamine receptor binding
beta-adrenergic receptor kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header