This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TOR1A SIRNA Assays ›
Gene Symbol:
Tor1a
Gene Name:
torsin family 1, member A (torsin A)
Gene Aliases:
DQ2, Dyt1, torsinA
Chromosome Location:
Chr.2: 30960561 - 30967918 on Build GRCm38
UniGene ID:
Mm.154994
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS241383
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tor1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144884.2 NP_659133.1 2 494
GenBank mRNA AJ298841.1 - 2 449
AK038891.1 - 2 452
AK079728.1 - 1 766
AK082936.1 - 2 404
AK160361.1 - 2 450
BC017683.1 - 2 474 4238480
BY724297.1 - 2 483

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein deneddylation
response to oxidative stress
organelle organization
nuclear envelope organization
cell adhesion
neuron projection development
protein localization to nucleus
wound healing, spreading of cells
intermediate filament cytoskeleton organization
synaptic vesicle transport
chaperone mediated protein folding requiring cofactor
protein homooligomerization
regulation of dopamine uptake involved in synaptic transmission
chaperone-mediated protein folding
ER-associated misfolded protein catabolic process
nuclear membrane organization
chaperone-mediated protein transport
positive regulation of synaptic vesicle endocytosis
regulation of protein localization to cell surface
nucleotide binding
protein binding
ATP binding
cytoskeletal protein binding
hydrolase activity
ATPase activity
kinesin binding
unfolded protein binding
misfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header