This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYN3 SIRNA Assays ›
Gene Symbol:
Syn3
Gene Name:
synapsin III
Gene Aliases:
-
Chromosome Location:
Chr.10: 86048746 - 86499019 on Build GRCm38
UniGene ID:
Mm.394931
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS241145
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164495.1 NP_001157967.1 4 683
NM_013722.3 NP_038750.2 4 683
GenBank mRNA AF192754.1 - 4 628
AF498252.1 - 3 430
AF498253.1 - 3 430
AK043854.1 - 4 684
AK048643.1 - 4 721
AK080557.1 - 4 644
AK082445.1 - 4 721
AK084032.1 - 4 669
AK086955.1 - 630
AK087342.1 - 5 759
BC107397.2 - 3 549 40058225

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017109, AF056704.

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter secretion
nucleotide binding
catalytic activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0