This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AHCYL1 SIRNA Assays ›
Gene Symbol:
Ahcyl1
Gene Name:
S-adenosylhomocysteine hydrolase-like 1
Gene Aliases:
1110034F20Rik, AA409031, AA414901, Ahcy-rs3, DCAL, Irbit
Chromosome Location:
Chr.3: 107663120 - 107696548 on Build GRCm38
UniGene ID:
Mm.220328
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS239005
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ahcyl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145542.3 NP_663517.2 14, 15 1648
GenBank mRNA AB092504.1 - 14, 15 1377
AK078823.1 - 7, 8 850
BC018218.1 - 14, 15 1477 4007102

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108561, XM_006233165, XM_008761406, XM_008761405, XM_006233167, and others.

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
protein export from nucleus
one-carbon metabolic process
positive regulation of sodium ion transport
regulation of mRNA 3'-end processing
regulation of ion transmembrane transporter activity
S-adenosylmethionine cycle
angiotensin-activated signaling pathway
regulation of anion transport
response to calcium ion
adenosylhomocysteinase activity
protein binding
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0