This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VIP SIRNA Assays ›
Gene Symbol:
Vip
Gene Name:
vasoactive intestinal polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.10: 5639193 - 5647617 on Build GRCm38
UniGene ID:
Mm.98916
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS238659
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011702.2 NP_035832.1 5 613
GenBank mRNA AK018599.1 - 5 614
AK043544.1 - 4 2334
BC089511.1 - 5 519 30249277

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endothelial cell proliferation
learning or memory
regulation of signal transduction
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
regulation of protein localization
negative regulation of apoptotic process
negative regulation of potassium ion transport
positive regulation of protein catabolic process
positive regulation of vasodilation
negative regulation of smooth muscle cell proliferation
regulation of sensory perception of pain
positive regulation of penile erection
receptor binding
hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header