This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SDF4 SIRNA Assays ›
Gene Symbol:
Sdf4
Gene Name:
stromal cell derived factor 4
Gene Aliases:
Cab45
Chromosome Location:
Chr.4: 155992843 - 156013610 on Build GRCm38
UniGene ID:
Mm.293517
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS237858
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sdf4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK014418.1 - 2 492
AK157860.1 - 2 357
AK163400.1 - 2 477
AK167113.1 - 2 406
BC005715.1 - 3 626
BC068152.1 - 2 732 30534982
D50461.1 - 2 735
U45977.1 - 2 640
U45978.1 - 2 508

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016547, NM_016176, BC011244, BC007625, AK075352, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0