This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPRF SIRNA Assays ›
Gene Symbol:
Ptprf
Gene Name:
protein tyrosine phosphatase, receptor type, F
Gene Aliases:
AA591035, LAR, LARS, RPTP-LAR
Chromosome Location:
Chr.4: 118208213 - 118291400 on Build GRCm38
UniGene ID:
Mm.29855
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS237675
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptprf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011213.2 NP_035343.2 8 1126
GenBank mRNA AF300943.1 - 6 789
AI385919.1 - 4 507
CD352108.1 - 1 52

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine-mediated signaling pathway
protein dephosphorylation
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
signal transduction
nervous system development
negative regulation of cell proliferation
regulation of neuron projection development
dephosphorylation
cell migration
neuron projection regeneration
negative regulation of cell projection organization
peptidyl-tyrosine dephosphorylation
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of insulin receptor signaling pathway
regulation of axon regeneration
positive regulation of dendrite morphogenesis
regulation of synapse structure or activity
negative regulation of neurotrophin TRK receptor signaling pathway
negative regulation of receptor binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
insulin receptor binding
heparin binding
hydrolase activity
phosphatase activity
receptor tyrosine kinase binding
protein complex binding
chondroitin sulfate proteoglycan binding
phosphate ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header