This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VMN1R21 SIRNA Assays ›
Gene Symbol:
Vmn1r21
Gene Name:
vomeronasal 1 receptor 21
Gene Aliases:
V1rc28
Chromosome Location:
Chr.6: 57843564 - 57844457 on Build GRCm38
UniGene ID:
Mm.377179
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS237174
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vmn1r21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134183.1 NP_598944.1 1 469
GenBank mRNA BC120799.1 - 1 510 40129722
BC120801.1 - 1 510 40129724

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
response to pheromone
pheromone binding
pheromone receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0