This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LGALS6 SIRNA Assays ›
Gene Symbol:
Lgals6
Gene Name:
lectin, galactose binding, soluble 6
Gene Aliases:
galectin-6
Chromosome Location:
-
UniGene ID:
Mm.210336
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS237047
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lgals6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010707.2 NP_034837.1 674
GenBank mRNA AF026794.1 - 686
EF017942.1 - 1001

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Process(es)

carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0