This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HS3ST3A1 SIRNA Assays ›
Gene Symbol:
Hs3st3a1
Gene Name:
heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1
Gene Aliases:
3Ost3a, Hs3st3a
Chromosome Location:
Chr.11: 64435332 - 64522835 on Build GRCm38
UniGene ID:
Mm.340214
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS236811
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hs3st3a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178870.5 NP_849201.1 2 1397
GenBank mRNA AK032282.1 - 2 365
AK051284.1 - 2 1398
BC094320.1 - 2 1451 6400013

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
sulfotransferase activity
[heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header