This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other S1PR3 SIRNA Assays ›
Gene Symbol:
S1pr3
Gene Name:
sphingosine-1-phosphate receptor 3
Gene Aliases:
AI132464, Edg3, Lpb3, S1p3
Chromosome Location:
Chr.13: 51408618 - 51422797 on Build GRCm38
UniGene ID:
Mm.136736
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS236378
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: S1pr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010101.4 NP_034231.1 2 1116
GenBank mRNA AB028143.1 - 2 984
AK028043.1 - 2 973
AK029852.1 - 2 972
AK030134.1 - 2 971
AK047268.1 - 2 964
AK078443.1 - 2 970
AK081919.1 - 2 961
AK083405.1 - 2 950
AK084944.1 - 2 971
AK085180.1 - 2 971
AK087076.1 - 2 971
AK087139.1 - 2 970
BC064007.1 - 2 956 30244496
BC068176.1 - 2 956 30613259

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
Notch signaling pathway
regulation of interleukin-1 beta production
negative regulation of establishment of endothelial barrier
signal transducer activity
G-protein coupled receptor activity
integrin binding
sphingosine-1-phosphate receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0