This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTYH2 SIRNA Assays ›
Gene Symbol:
Ttyh2
Gene Name:
tweety homolog 2 (Drosophila)
Gene Aliases:
1110001A03Rik
Chromosome Location:
Chr.11: 114675468 - 114720984 on Build GRCm38
UniGene ID:
Mm.271934
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235923
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttyh2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053273.2 NP_444503.2 8 976
GenBank mRNA AF329682.1 - 8 919
AK152172.1 - 8 928
AK154835.1 - 8 984
AK155166.1 - 8 977
AK156393.1 - 8 929
AK168408.1 - 8 929
AK204126.1 - 1 18
BC060740.1 - 8 919 4158813
BY731753.1 - 4 287

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
biological_process
molecular_function
chloride channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0