This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TECR SIRNA Assays ›
Gene Symbol:
Tecr
Gene Name:
trans-2,3-enoyl-CoA reductase
Gene Aliases:
2410016D23Rik, A230102P12Rik, AI173355, D17Ertd178e, Gpsn2, SC2
Chromosome Location:
Chr.8: 83571698 - 83594491 on Build GRCm38
UniGene ID:
Mm.352239
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235569
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tecr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027179.1 NP_081455.1 7 609
NM_134118.5 NP_598879.1 8 654
GenBank mRNA AK002384.1 - 8 624
AK010513.1 - 7 545
AK010984.1 - 8 628
AK013558.1 - 8 607
AK171718.1 - 3 361
AK172506.1 - 3 339
AK189199.1 - 4 185
BC019984.1 - 8 640 4240045
BC094046.1 - 8 754 5064002

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
steroid biosynthetic process
fatty acid elongation
very long-chain fatty acid biosynthetic process
oxidation-reduction process
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0