This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DDX50 SIRNA Assays ›
Gene Symbol:
Ddx50
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 50
Gene Aliases:
4933429B04Rik, 8430408E17Rik, GU2, RH-II
Chromosome Location:
Chr.10: 62616015 - 62651606 on Build GRCm38
UniGene ID:
Mm.114116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235113
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddx50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053183.2 NP_444413.1 13 1950
GenBank mRNA AF334104.1 - 13 1951
AK030239.1 - 14 2022
AK041281.1 - 13 1849
BC049906.1 - 13 1951
BC055440.1 - 13 1945
BC075714.1 - 13 1903
BC090996.1 - 13 1970 6757408

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_024045, BC000272, BC000210, AK293577, AF334103.
This siRNA also targets the following transcripts in Rat: NM_001013198, BC091427.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0