This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OPHN1 SIRNA Assays ›
Gene Symbol:
Ophn1
Gene Name:
oligophrenin 1
Gene Aliases:
C130037N19Rik
Chromosome Location:
Chr.X: 98554280 - 98891414 on Build GRCm38
UniGene ID:
Mm.254336
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235104
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ophn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052976.3 NP_443208.1 2 688
GenBank mRNA AK028822.1 - 2 172
AK031419.1 - 2 681
AK032500.1 - 2 330
AK036038.1 - 2 689
AK039304.1 - 1 856
AK043239.1 - 2 689
AK046214.1 - 2 144
AK048153.1 - 2 689
AK077982.1 - 2 313
AK082551.1 - 2 328
AK087469.1 - 2 152
BC004845.1 - 3 286 2650009

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endocytosis
signal transduction
nervous system development
actin cytoskeleton organization
regulation of endocytosis
positive regulation of GTPase activity
synaptic vesicle endocytosis
regulation of synaptic transmission, glutamatergic
negative regulation of proteasomal protein catabolic process
actin binding
GTPase activator activity
protein binding
phospholipid binding
ionotropic glutamate receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0