This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TNFRSF21 SIRNA Assays ›
Gene Symbol:
Tnfrsf21
Gene Name:
tumor necrosis factor receptor superfamily, member 21
Gene Aliases:
AA959878, DR6, R74815, TR7
Chromosome Location:
Chr.17: 43016555 - 43089188 on Build GRCm38
UniGene ID:
Mm.200792
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235094
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tnfrsf21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178589.3 NP_848704.1 5 2157
GenBank mRNA AF322069.1 - 5 1715
AK033529.1 - 5 2133
AK043823.1 - 5 2159
AK134704.1 - 5 2128
AY043489.1 - 5 1715
BC016420.1 - 5 2106 4220624

Back To Top

More Information


Gene Ontology Categories:

Function(s)

B cell apoptotic process
adaptive immune response
immune system process
apoptotic process
humoral immune response
signal transduction
negative regulation of B cell proliferation
negative regulation of myelination
negative regulation of T cell proliferation
myelination
regulation of oligodendrocyte differentiation
T cell receptor signaling pathway
neuron apoptotic process
cellular response to tumor necrosis factor
oligodendrocyte apoptotic process
negative regulation of interleukin-5 secretion
negative regulation of interleukin-13 secretion
negative regulation of interleukin-10 secretion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0