This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ICE2 SIRNA Assays ›
Gene Symbol:
Ice2
Gene Name:
interactor of little elongation complex ELL subunit 2
Gene Aliases:
B230343B06Rik, Narg2
Chromosome Location:
Chr.9: 69397911 - 69433076 on Build GRCm38
UniGene ID:
Mm.35911
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS234874
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ice2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145618.3 NP_663593.2 9 1134
GenBank mRNA AK134186.1 - 9 1179
AK163917.1 - 9 1062
AY244558.1 - 9 1163
BC068183.1 - 9 1134 30639735

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header