This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEC16B SIRNA Assays ›
Gene Symbol:
Sec16b
Gene Name:
SEC16 homolog B (S. cerevisiae)
Gene Aliases:
Lztr2, Rgpr, Rgpr-p117
Chromosome Location:
Chr.1: 157506733 - 157568425 on Build GRCm38
UniGene ID:
Mm.87114
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS234835
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sec16b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159986.1 NP_001153458.1 14, 15 1862
NM_033354.3 NP_203505.3 16, 17 2109
GenBank mRNA AB063356.1 - 14, 15 1862
AK132144.1 - 14, 15 1886
AK173302.1 - 16, 17 2090
BC039223.1 - 6, 7 713
BC051110.1 - 7, 8 809
BC059194.1 - 15, 16 1927 4222915

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
endoplasmic reticulum organization
peroxisome organization
positive regulation of gene expression
protein transport
vesicle-mediated transport
peroxisome fission
COPII vesicle coating
positive regulation of protein exit from endoplasmic reticulum
protein localization to endoplasmic reticulum
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0