This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM2 SIRNA Assays ›
Gene Symbol:
Trim2
Gene Name:
tripartite motif-containing 2
Gene Aliases:
mKIAA0517, narf
Chromosome Location:
Chr.3: 84160439 - 84305832 on Build GRCm38
UniGene ID:
Mm.44876
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS234559
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271725.1 NP_001258654.1 3 352
NM_001271726.1 NP_001258655.1 3 388
NM_001271727.1 NP_001258656.1 3 657
NM_001271728.1 NP_001258657.1 3 455
NM_030706.3 NP_109631.2 3 451
GenBank mRNA AB043550.1 - 3 435
AF220017.1 - 3 300
AK039741.1 - 3 449
AK045410.1 - 3 448
AK046887.1 - 3 436
AK079415.1 - 3 602
AK083057.1 - 2 2210
AK141419.1 - 3 298
AK147275.1 - 3 565
AK147395.1 - 3 446
AK164350.1 - 2 154
AK220343.1 - 3 656
BC058668.1 - 3 419
BC058961.1 - 3 455 6826143
CD802800.1 - 3 314
CD803806.1 - 3 451
CJ100776.1 - 3 353

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of neuron apoptotic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
myosin binding
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0