This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC24A2 SIRNA Assays ›
Gene Symbol:
Slc24a2
Gene Name:
solute carrier family 24 (sodium/potassium/calcium exchanger), member 2
Gene Aliases:
2810021B17Rik, 6330417K15Rik, AI847460, Nckx2
Chromosome Location:
Chr.4: 86983124 - 87230554 on Build GRCm38
UniGene ID:
Mm.490487
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS233540
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc24a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110240.1 NP_001103710.1 1 636
NM_172426.2 NP_766014.1 2 861
GenBank mRNA AK034424.1 - 2 745
AK038634.1 - 2 745
AK038800.1 - 2 789
AK083127.1 - 2 745
AK136058.1 - 2 790
BC115867.1 - 1 638 40081382
BC116637.1 - 1 638 40081381

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031743, AF021923.

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
cellular calcium ion homeostasis
learning
memory
ion transmembrane transport
sodium ion transmembrane transport
long-term synaptic potentiation
long term synaptic depression
calcium ion transmembrane transport
potassium ion transmembrane transport
calcium channel activity
calcium ion binding
calcium, potassium:sodium antiporter activity
nickel cation binding
manganese ion binding
potassium ion binding
sodium ion binding
cadmium ion binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0