This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB11FIP1 SIRNA Assays ›
Gene Symbol:
Rab11fip1
Gene Name:
RAB11 family interacting protein 1 (class I)
Gene Aliases:
2010200K21Rik, 4833414G05Rik, Rcp
Chromosome Location:
Chr.8: 27138773 - 27174646 on Build GRCm38
UniGene ID:
Mm.205335
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS233275
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab11fip1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080813.2 NP_001074282.1 2 568
NM_029423.2 NP_083699.2 2 568
GenBank mRNA AK014696.1 - 2 550
AK030769.1 - 2 571
AK086797.1 - 2 566
AK151024.1 - 2 569
BC125400.1 - 2 553 40129915
BC132094.1 - 2 553 40130371

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
regulated exocytosis
negative regulation of adiponectin secretion
molecular_function
Rab GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0