This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLF17 SIRNA Assays ›
Gene Symbol:
Klf17
Gene Name:
Kruppel-like factor 17
Gene Aliases:
7420700M05Rik, AA420409, AU043468, C85123, D4Ertd561e, Gzf, Zfp393
Chromosome Location:
Chr.4: 117756659 - 117765666 on Build GRCm38
UniGene ID:
Mm.3848
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS233266
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klf17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029416.2 NP_083692.2 2 456
GenBank mRNA AF508984.1 - 2 422
AK018361.1 - 2 422
AK139827.1 - 2 423
BC064748.1 - 2 400 30476790

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
gamete generation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0