This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZKSCAN1 SIRNA Assays ›
Gene Symbol:
Zkscan1
Gene Name:
zinc finger with KRAB and SCAN domains 1
Gene Aliases:
5930429A01Rik, 9130423L19Rik, 9230118B16Rik, AI646829, AI788588, C87323, KOX18, PHZ-37, ZNF139
Chromosome Location:
Chr.5: 138085076 - 138107822 on Build GRCm38
UniGene ID:
Mm.213114
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232763
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zkscan1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029869.1 NP_084145.1 4 1075
NM_133906.4 NP_598667.2 6 1300
GenBank mRNA AK028932.1 - 6 1231
AK031759.1 - 6 1281
AK036807.1 - 6 1283
AK054323.1 - 6 1271
AK131148.1 - 3 1220
AK135999.1 - 6 1250
AK154831.1 - 6 1280
BC052441.1 - 4 1039 6406518

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0