This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OSBPL10 SIRNA Assays ›
Gene Symbol:
Osbpl10
Gene Name:
oxysterol binding protein-like 10
Gene Aliases:
4933433D06Rik, C820004B04Rik, OPR-10
Chromosome Location:
Chr.9: 114978583 - 115233273 on Build GRCm38
UniGene ID:
Mm.458474
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232714
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Osbpl10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148958.2 NP_683761.1 6 1043
GenBank mRNA AK017034.1 - 8 1374
AK037617.1 - 5 1185
BC027256.1 - 6 1043 2650570

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
cholesterol binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header