This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAP10 SIRNA Assays ›
Gene Symbol:
Map10
Gene Name:
microtubule-associated protein 10
Gene Aliases:
4933403G14Rik, AI835009, mKIAA1383
Chromosome Location:
Chr.8: 125669818 - 125673365 on Build GRCm38
UniGene ID:
Mm.41709
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232614
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Map10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028908.3 NP_083184.1 1 1710
GenBank mRNA AK016632.1 - 1 1666
AK042358.1 - 1 1683
AK080043.1 - 1 1664
AK220388.1 - 1 1637

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
cell cycle
cytoplasmic microtubule organization
positive regulation of cytokinesis
regulation of microtubule-based process
mitotic spindle midzone assembly
cell division
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0