This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTF2 SIRNA Assays ›
Gene Symbol:
Ttf2
Gene Name:
transcription termination factor, RNA polymerase II
Gene Aliases:
4632434F22Rik, 8030447N19, AV218430
Chromosome Location:
Chr.3: 100938860 - 100969725 on Build GRCm38
UniGene ID:
Mm.270961
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232247
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013026.2 NP_001013044.2 11 1951
GenBank mRNA AK014607.1 - 11 1939
AK078759.1 - 11 1951
AK087352.1 - 11 1951
BC087733.1 - 11 1958 30434981
BC096625.1 - 11 1949 30109731

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
DNA-templated transcription, termination
regulation of transcription, DNA-templated
mRNA processing
biological_process
RNA splicing
nucleotide binding
nucleic acid binding
DNA binding
helicase activity
ATP binding
ATP-dependent helicase activity
zinc ion binding
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header