This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC30 SIRNA Assays ›
Gene Symbol:
Ccdc30
Gene Name:
coiled-coil domain containing 30
Gene Aliases:
1700001O02Rik, 1700041C02Rik, 1700111D19Rik, 4930445I03Rik
Chromosome Location:
Chr.4: 119322893 - 119415521 on Build GRCm38
UniGene ID:
Mm.425204
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS231980
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc30
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270435.1 NP_001257364.1 2, 3 585
NM_029286.2 NP_083562.1 2, 3 585
GenBank mRNA AK078362.1 - 2, 3 512
AK148014.1 - 2, 3 568
AK163185.1 - 2, 3 526
AK225009.1 - 2, 3 544
BC114436.1 - 2, 3 512 40059415

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header