This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC56 SIRNA Assays ›
Gene Symbol:
Lrrc56
Gene Name:
leucine rich repeat containing 56
Gene Aliases:
5730427C23Rik, BB110509, mFLJ00101
Chromosome Location:
Chr.7: 141194116 - 141210055 on Build GRCm38
UniGene ID:
Mm.45104
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS230285
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc56
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172064.1 NP_001165535.1 8 1215
NM_001172065.1 NP_001165536.1 8 1215
NM_153777.2 NP_722472.1 8 801
GenBank mRNA AK017603.1 - 8 1189
AK220237.1 - 8 1169
BC027807.1 - 8 811 5324022

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0