This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KPTN SIRNA Assays ›
Gene Symbol:
Kptn
Gene Name:
kaptin
Gene Aliases:
2310042D10Rik, 2E4, C030013F01Rik
Chromosome Location:
Chr.7: 16119876 - 16127516 on Build GRCm38
UniGene ID:
Mm.41101
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS230193
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kptn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133727.2 NP_598488.2 1 286
GenBank mRNA AK009746.1 - 1 267
AK081226.1 - 1 278
BC018316.1 - 1 254 4160465
BY269246.1 - 1 288

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
biological_process
molecular_function
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0