This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP4R1 SIRNA Assays ›
Gene Symbol:
Ppp4r1
Gene Name:
protein phosphatase 4, regulatory subunit 1
Gene Aliases:
3110001J10Rik, Pp4r1
Chromosome Location:
Chr.17: 65782398 - 65841926 on Build GRCm38
UniGene ID:
Mm.48686
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS230155
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp4r1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114131.1 NP_001107603.1 15 2104
NM_146081.2 NP_666193.2 15 2155
GenBank mRNA AK013951.2 - 2 177
AK133969.1 - 15 2027
AK166418.1 - 15 2062
AK167713.1 - 14 1566
BC020371.1 - 3 239
BC026489.1 - 7 1128 4504305
BC029761.1 - 15 2090 3989055

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0