This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GTF3C5 SIRNA Assays ›
Gene Symbol:
Gtf3c5
Gene Name:
general transcription factor IIIC, polypeptide 5
Gene Aliases:
2700084A09Rik, TFIIIC63, TFiiiC2-63
Chromosome Location:
Chr.2: 28566245 - 28583746 on Build GRCm38
UniGene ID:
Mm.274120
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS230118
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf3c5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290484.1 NP_001277413.1 2 476
NM_148928.3 NP_683730.1 2 476
GenBank mRNA AK045419.1 - 2 68
AK137359.1 - 2 465
AK147014.1 - 2 454
BC027055.1 - 2 443 5360397
BC027247.1 - 2 386 2631223
BC039171.1 - 2 397 4500799
BQ179286.1 - 2 476

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
skeletal muscle cell differentiation
molecular_function
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0