This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NKIRAS1 SIRNA Assays ›
Gene Symbol:
Nkiras1
Gene Name:
NFKB inhibitor interacting Ras-like protein 1
Gene Aliases:
2400004O09Rik, KBRAS1, kappaB-Ras1
Chromosome Location:
Chr.14: 18271142 - 18284003 on Build GRCm38
UniGene ID:
Mm.25648
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229855
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nkiras1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023526.3 NP_076015.2 3 491
GenBank mRNA AF225707.1 - 3 348
AK010285.1 - 4 664
AK028539.1 - 3 479
AK148040.1 - 3 417
AK167340.1 - 3 388
BC061092.1 - 3 396 6589347

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
small GTPase mediated signal transduction
biological_process
nucleotide binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0