This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK13 SIRNA Assays ›
Gene Symbol:
Cdk13
Gene Name:
cyclin-dependent kinase 13
Gene Aliases:
2310015O17Rik, Cdc2l5
Chromosome Location:
Chr.13: 17715962 - 17805332 on Build GRCm38
UniGene ID:
Mm.193924
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229729
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081058.2 NP_001074527.1 2 2079
NM_027118.1 NP_081394.1 2 2079
GenBank mRNA AK035493.1 - 2 753
AK083577.1 - 2 465
AK173265.1 - 2 2079
BC076619.1 - 3 534

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
mRNA processing
protein phosphorylation
RNA splicing
phosphorylation
hemopoiesis
phosphorylation of RNA polymerase II C-terminal domain
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
cyclin binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0