This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM45A2 SIRNA Assays ›
Gene Symbol:
Tmem45a2
Gene Name:
transmembrane protein 45A2
Gene Aliases:
2310005G13Rik
Chromosome Location:
Chr.16: 57036967 - 57072092 on Build GRCm38
UniGene ID:
Mm.58105
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229683
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem45a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183281.2 NP_899104.2 3 269
GenBank mRNA AK009173.1 - 3 277
AK036849.1 - 3 296
BC150914.1 - 2 422
BC150924.1 - 2 422
BC172084.1 - 2 422 9054370

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0