This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 9530077C05RIK SIRNA Assays ›
Gene Symbol:
9530077C05Rik
Gene Name:
RIKEN cDNA 9530077C05 gene
Gene Aliases:
1110003N12Rik, Kiaa0895, mKIAA0895
Chromosome Location:
Chr.9: 22387732 - 22444684 on Build GRCm38
UniGene ID:
Mm.280669
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229034
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 9530077C05Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026739.1 NP_081015.1 3 933
GenBank mRNA AK220359.1 - 3 997
BC039998.1 - 1 156 3326047
BC054761.1 - 3 933 6413638

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008766018, XM_235936, XM_006242702, XM_006242701, FQ217109.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0