This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM9SF2 SIRNA Assays ›
Gene Symbol:
Tm9sf2
Gene Name:
transmembrane 9 superfamily member 2
Gene Aliases:
1500001N15Rik, AA536814, D14Ertd64e, P76
Chromosome Location:
Chr.14: 122107082 - 122159603 on Build GRCm38
UniGene ID:
Mm.275191
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228895
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm9sf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080556.3 NP_542123.3 9 1112
GenBank mRNA AK010325.1 - 9 1069
AK030340.1 - 9 1043
AK048012.1 - 9 1031
AK050338.1 - 9 1059
AK075720.1 - 9 1112
AK088905.1 - 9 1084
AK159757.1 - 9 1057
BC003862.1 - 9 1051 3488326

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0