This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPA1 SIRNA Assays ›
Gene Symbol:
Ppa1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
2010317E03Rik, Pyp
Chromosome Location:
Chr.10: 61648621 - 61674165 on Build GRCm38
UniGene ID:
Mm.28897
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228755
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppa1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026438.4 NP_080714.2 8, 9 823
GenBank mRNA AK008575.1 - 8, 9 824
AK020291.1 - 8, 9 839
AK088397.1 - 8, 9 819
AK145640.1 - 8, 9 873
AK146300.1 - 8, 9 786
AK151512.1 - 8, 9 814
AK201001.1 - 2, 3 177
AK216005.1 - 2, 3 177
BC010468.1 - 8, 9 757 3585780
CT010203.1 - 8, 9 716

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
magnesium ion binding
inorganic diphosphatase activity
pyrophosphatase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0