This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGARP SIRNA Assays ›
Gene Symbol:
Mgarp
Gene Name:
mitochondria localized glutamic acid rich protein
Gene Aliases:
4930583H14Rik, AI195347, CESP-1, HUMMR, Osap, Qsap
Chromosome Location:
Chr.3: 51388412 - 51396547 on Build GRCm38
UniGene ID:
Mm.273339
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228638
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mgarp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026358.3 NP_080634.2 4 388
GenBank mRNA AF228502.1 - 4 367
AK006339.1 - 4 350
AK016342.1 - 4 361
AK151401.1 - 4 382
BC012511.1 - 4 381 4500810
BC022740.1 - 4 542 4511440
BC049556.1 - 4 350 6772062

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to mitochondrion
anterograde axonal transport
retrograde axonal transport
positive regulation of mitochondrion organization
axonal transport of mitochondrion
cellular response to steroid hormone stimulus
cellular response to hypoxia
cellular response to gonadotropin-releasing hormone
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0