This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SDHB SIRNA Assays ›
Gene Symbol:
Sdhb
Gene Name:
succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
Gene Aliases:
0710008N11Rik
Chromosome Location:
Chr.4: 140961271 - 140979197 on Build GRCm38
UniGene ID:
Mm.246965
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228563
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sdhb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023374.3 NP_075863.2 8 934
GenBank mRNA AK003052.1 - 8 828
AK003533.1 - 8 818
AK009660.1 - 8 828
AK153045.1 - 8 811
AK169252.1 - 8 825
AK169839.1 - 8 827
BC051934.1 - 8 934 5688898

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
succinate metabolic process
transport
respiratory electron transport chain
oxidation-reduction process
succinate dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
3 iron, 4 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0