This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOXL4 SIRNA Assays ›
Gene Symbol:
Loxl4
Gene Name:
lysyl oxidase-like 4
Gene Aliases:
4833426I20Rik, Loxc
Chromosome Location:
Chr.19: 42592279 - 42612906 on Build GRCm38
UniGene ID:
Mm.178767
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228497
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Loxl4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164311.1 NP_001157783.1 8 1300
NM_053083.3 NP_444313.3 8 1297
GenBank mRNA AF338440.1 - 7 1192
AK014331.1 - 8 1300
BC068307.1 - 8 1239 30059745

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
protein-lysine 6-oxidase activity
scavenger receptor activity
copper ion binding
aspartate oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor
D-lysine oxidase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header