This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LARP6 SIRNA Assays ›
Gene Symbol:
Larp6
Gene Name:
La ribonucleoprotein domain family, member 6
Gene Aliases:
5430431G03Rik, AI552438, Achn
Chromosome Location:
Chr.9: 60713121 - 60738801 on Build GRCm38
UniGene ID:
Mm.44546
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228488
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Larp6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026235.4 NP_080511.2 3 1036
GenBank mRNA AK017372.1 - 3 1041
AK035809.1 - 613
AK039614.1 - 3 1019
AK047452.1 - 1 340
AK087521.1 - 3 1039
BC090615.1 - 3 1168 6809572

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

RNA processing
regulation of translation
nucleotide binding
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0