This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZC3H13 SIRNA Assays ›
Gene Symbol:
Zc3h13
Gene Name:
zinc finger CCCH type containing 13
Gene Aliases:
2600010B19Rik, 3110050K21Rik, 4930570G11Rik, C87618
Chromosome Location:
Chr.14: 75284308 - 75344433 on Build GRCm38
UniGene ID:
Mm.74704
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228224
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zc3h13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026083.2 NP_080359.2 4, 5 631
GenBank mRNA AK014200.1 - 4, 5 411
AK078225.1 - 4, 5 622
BC138264.1 - 4, 5 580 8861283

Back To Top

More Information


Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0