This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTGR1 SIRNA Assays ›
Gene Symbol:
Ptgr1
Gene Name:
prostaglandin reductase 1
Gene Aliases:
2510002C21Rik, Ltb4dh, Zadh3
Chromosome Location:
Chr.4: 58965590 - 58987131 on Build GRCm38
UniGene ID:
Mm.34497
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228020
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptgr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025968.3 NP_080244.1 5 304
GenBank mRNA AK010888.1 - 5 339
AK011962.1 - 5 326
AK035425.1 - 5 305
AK134440.1 - 5 337
AK166835.1 - 5 304
BC014865.1 - 5 286 2648051
CT010267.1 - 4 253

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
zinc ion binding
oxidoreductase activity
enoyl-[acyl-carrier-protein] reductase activity
2,3-dihydroxy-2,3-dihydro-phenylpropionate dehydrogenase activity
cis-2,3-dihydrodiol DDT dehydrogenase activity
trans-9R,10R-dihydrodiolphenanthrene dehydrogenase activity
cis-chlorobenzene dihydrodiol dehydrogenase activity
2,5-dichloro-2,5-cyclohexadiene-1,4-diol dehydrogenase activity
trans-1,2-dihydrodiolphenanthrene dehydrogenase activity
2-alkenal reductase [NAD(P)] activity
3,4-dihydroxy-3,4-dihydrofluorene dehydrogenase activity
benzo(a)pyrene-trans-11,12-dihydrodiol dehydrogenase activity
benzo(a)pyrene-cis-4,5-dihydrodiol dehydrogenase activity
citronellyl-CoA dehydrogenase activity
menthone dehydrogenase activity
phthalate 3,4-cis-dihydrodiol dehydrogenase activity
13-prostaglandin reductase activity
cinnamate reductase activity
15-oxoprostaglandin 13-oxidase activity
NADPH-dependent curcumin reductase activity
NADPH-dependent dihydrocurcumin reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0