This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LONP2 SIRNA Assays ›
Gene Symbol:
Lonp2
Gene Name:
lon peptidase 2, peroxisomal
Gene Aliases:
1300002A08Rik, AU015403, Lonp
Chromosome Location:
Chr.8: 86624043 - 86716636 on Build GRCm38
UniGene ID:
Mm.324550
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227833
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lonp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168591.1 NP_001162063.1 6 705
NM_025827.3 NP_080103.1 12 2009
GenBank mRNA AK004843.1 - 12 1923
AK050231.1 - 12 1918
AK075719.1 - 6 706
AK088348.1 - 11 1915
AK151103.1 - 12 1955
AK151406.1 - 10 1523
AK152964.1 - 10 1524
AK168628.1 - 10 1533
AK168791.1 - 12 1955
AK168956.1 - 12 1955
AK169656.1 - 1923
BC049090.1 - 12 1945 6401542

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
misfolded or incompletely synthesized protein catabolic process
protein targeting to peroxisome
protein processing
protein catabolic process
regulation of fatty acid beta-oxidation
nucleotide binding
protease binding
ATP-dependent peptidase activity
serine-type endopeptidase activity
receptor binding
protein binding
ATP binding
peptidase activity
serine-type peptidase activity
hydrolase activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0