This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2W SIRNA Assays ›
Gene Symbol:
Ube2w
Gene Name:
ubiquitin-conjugating enzyme E2W (putative)
Gene Aliases:
6130401J04Rik
Chromosome Location:
Chr.1: 16540788 - 16619420 on Build GRCm38
UniGene ID:
Mm.122430
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227737
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2w
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271016.1 NP_001257945.1 1, 2 101
NM_001271017.1 NP_001257946.1 1, 2 101
NM_025773.3 NP_080049.2 1, 2 101
GenBank mRNA AK015348.1 - 1, 2 55
AK042870.1 - 1, 2 51
AK044463.1 - 1, 2 53
AK046086.1 - 1, 2 55
AK050679.1 - 1, 2 53
AK050923.1 - 1, 2 75
AK077997.1 - 1, 2 26
AK082233.1 - 1, 2 50
BC020124.1 - 1, 2 53 4012785
BY728312.1 - 1, 2 101
CN663576.1 - 1, 2 21
DV044691.1 - 50

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008773483, XM_006237922, BC168682.

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
protein monoubiquitination
misfolded or incompletely synthesized protein catabolic process
cellular response to DNA damage stimulus
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K11-linked ubiquitination
cellular response to misfolded protein
nucleotide binding
ubiquitin-protein transferase activity
protein binding
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0